siRNA Details
| Gene Symbol: Hsd17b10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016763.2 | NP_058043.3 | 5 | 589 | ||
| GenBank mRNA | AF233685.1 | - | 5 | 589 | ||
| AK002368.1 | - | 5 | 584 | |||
| AK013340.1 | - | 5 | 585 | |||
| AK077735.1 | - | 5 | 583 | |||
| AK088685.1 | - | 5 | 605 | |||
| BC027517.1 | - | 5 | 589 | 3376098 | ||
| U96116.1 | - | 5 | 690 | |||

Hsd17b10
Genomic Map