siRNA Details
| Gene Symbol: Hand1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008213.2 | NP_032239.1 | 1 | 438 | ||
| GenBank mRNA | AK146084.1 | - | 339 | |||
| BC050182.1 | - | 1 | 288 | 30022934 | ||
| S79216.1 | - | 1 | 397 | |||
| U21226.1 | - | 1 | 438 | |||
| U43714.1 | - | 1 | 296 | |||

Hand1
Genomic Map