siRNA Details
| Gene Symbol: Hap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010404.3 | NP_034534.1 | 3, 4 | 647 | ||
| NM_177981.2 | NP_817090.1 | 3, 4 | 647 | |||
| GenBank mRNA | AJ000262.1 | - | 3, 4 | 593 | ||
| AJ002271.1 | - | 3, 4 | 593 | |||
| AK138436.1 | - | 594 | ||||
| BC034089.1 | - | 3, 4 | 585 | 4481735 | ||
| BC053043.1 | - | 3, 4 | 634 | 5694628 | ||

Hap1
Genomic Map