siRNA Details
| Gene Symbol: DVL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004423.3 | NP_004414.3 | 8, 9 | 1051 | ||
| XM_005247172.2 | XP_005247229.1 | 8, 9 | 1236 | |||
| XM_011512513.2 | XP_011510815.1 | 5, 6 | 839 | |||
| GenBank mRNA | AF006013.1 | - | 8, 9 | 922 | ||
| AK093469.1 | - | 8, 9 | 1043 | |||
| AK304686.1 | - | 9, 10 | 1003 | |||
| AL110278.1 | - | 4, 5 | 693 | |||
| BC018289.1 | - | 6, 7 | 587 | |||
| BC032459.1 | - | 8, 9 | 979 | 5200610 | ||
| D86963.1 | - | 8, 9 | 1051 | |||
| U48252.1 | - | 1, 2 | 156 | |||
| U75651.1 | - | 8, 9 | 1304 | |||

DVL3
Genomic Map