siRNA Details
| Gene Symbol: Hdac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008229.2 | NP_032255.2 | 2 | 320 | ||
| GenBank mRNA | AB221663.1 | - | 2 | 112 | ||
| AK051753.1 | - | 2 | 295 | |||
| AK165734.1 | - | 2 | 321 | |||
| BC138517.1 | - | 2 | 292 | 8861537 | ||
| U31758.1 | - | 2 | 280 | |||

Hdac2
Genomic Map