siRNA Details
| Gene Symbol: Hdac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010411.2 | NP_034541.2 | 3 | 343 | ||
| GenBank mRNA | AF074881.1 | - | 3 | 290 | ||
| AF098295.1 | - | 3 | 268 | |||
| AF125536.1 | - | 3 | 275 | |||
| AK145158.1 | - | 3 | 309 | |||
| BC139300.1 | - | 3 | 371 | 8862322 | ||
| BC139301.1 | - | 3 | 371 | 8862323 | ||
| BQ932476.1 | - | 3 | 355 | |||
| CJ166955.1 | - | 3 | 346 | |||

Hdac3
Genomic Map