siRNA Details
| Gene Symbol: Hebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013546.2 | NP_038574.2 | 4 | 637 | ||
| GenBank mRNA | AB013095.1 | - | 4 | 576 | ||
| AF117613.1 | - | 4 | 625 | |||
| AK008919.1 | - | 4 | 558 | |||
| BC012654.1 | - | 4 | 561 | 4216585 | ||
| CT010262.1 | - | 4 | 519 | |||

Hebp1
Genomic Map