siRNA Details
| Gene Symbol: Hells | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008234.3 | NP_032260.2 | 21 | 2452 | ||
| GenBank mRNA | AF155210.1 | - | 21 | 2452 | ||
| AK013266.1 | - | 21 | 2491 | |||
| AK147126.1 | - | 21 | 2483 | |||
| AK166636.1 | - | 21 | 2453 | |||
| BC020056.1 | - | 8 | 816 | |||
| BC090671.1 | - | 21 | 2495 | |||
| BC100394.1 | - | 21 | 2449 | 30480539 | ||
| U25691.2 | - | 21 | 2485 | |||

Hells
Genomic Map