siRNA Details
| Gene Symbol: Hells | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008234.3 | NP_032260.2 | 10 | 982 | ||
| GenBank mRNA | AF155210.1 | - | 10 | 982 | ||
| AK013266.1 | - | 10 | 1021 | |||
| AK147126.1 | - | 10 | 1013 | |||
| AK166636.1 | - | 10 | 982 | |||
| BC090671.1 | - | 10 | 1027 | |||
| BC100394.1 | - | 10 | 979 | 30480539 | ||
| U25691.2 | - | 10 | 1015 | |||

Hells
Genomic Map