siRNA Details
| Gene Symbol: Hells | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008234.3 | NP_032260.2 | 22 | 2522 | ||
| GenBank mRNA | AF155210.1 | - | 22 | 2522 | ||
| AK013266.1 | - | 22 | 2561 | |||
| AK147126.1 | - | 22 | 2553 | |||
| AK166636.1 | - | 22 | 2523 | |||
| BC020056.1 | - | 9 | 886 | |||
| BC090671.1 | - | 22 | 2565 | |||
| BC100394.1 | - | 22 | 2519 | 30480539 | ||
| U25691.2 | - | 22 | 2555 | |||

Hells
Genomic Map