siRNA Details
| Gene Symbol: Hexb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010422.2 | NP_034552.1 | 7 | 886 | ||
| GenBank mRNA | AK151149.1 | - | 7 | 823 | ||
| AK159134.1 | - | 7 | 839 | |||
| AK159178.1 | - | 7 | 860 | |||
| AK160560.1 | - | 7 | 855 | |||
| AK168825.1 | - | 7 | 860 | |||
| U07633.1 | - | 7 | 858 | |||
| Y00964.1 | - | 7 | 799 | |||

Hexb
Genomic Map