siRNA Details
| Gene Symbol: Foxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277290.1 | NP_001264219.1 | 3 | 291 | ||
| NM_008238.2 | NP_032264.1 | 3 | 294 | |||
| GenBank mRNA | BC108980.1 | - | 2 | 179 | 40047616 | |
| BC108981.1 | - | 2 | 182 | 40047617 | ||
| CJ056441.1 | - | 3 | 293 | |||
| X81593.1 | - | 3 | 244 | |||

Foxn1
Genomic Map