siRNA Details
| Gene Symbol: Hiat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008246.2 | NP_032272.2 | 2, 3 | 377 | ||
| GenBank mRNA | AK004779.1 | - | 2, 3 | 316 | ||
| AK135865.1 | - | 2, 3 | 198 | |||
| AK158838.1 | - | 2, 3 | 378 | |||
| AK169626.1 | - | 2, 3 | 317 | |||
| BC118047.1 | - | 2, 3 | 197 | 40106779 | ||
| D88315.1 | - | 2, 3 | 250 | |||

Hiat1
Genomic Map