siRNA Details
| Gene Symbol: Hmbs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013551.2 | NP_038579.2 | 1, 2 | 228 | ||
| GenBank mRNA | AK151020.1 | - | 1, 2 | 194 | ||
| AK153884.1 | - | 1, 2 | 161 | |||
| AK166734.1 | - | 1, 2 | 126 | |||
| AK167702.1 | - | 1, 2 | 187 | |||
| BC003861.1 | - | 1, 2 | 188 | 3487642 | ||
| BY786671.1 | - | 1, 2 | 229 | |||

Hmbs
Genomic Map