siRNA Details
| Gene Symbol: Hmmr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013552.2 | NP_038580.2 | 13 | 1735 | ||
| GenBank mRNA | AF031932.1 | - | 13 | 1720 | ||
| AF079222.1 | - | 14 | 1684 | |||
| AK143123.1 | - | 13 | 1735 | |||
| AK163410.1 | - | 13 | 1615 | |||
| BC021427.1 | - | 1859 | 5035341 | |||
| X64550.1 | - | 9 | 1348 | |||

Hmmr
Genomic Map