siRNA Details
| Gene Symbol: Hmx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010445.2 | NP_034575.1 | 2 | 1299 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hmx1| Gene Symbol: Hmx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010445.2 | NP_034575.1 | 2 | 1299 | ||