siRNA Details
| Gene Symbol: Hmx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145998.3 | NP_666110.1 | 2 | 1208 | ||
| GenBank mRNA | AK053014.1 | - | 2 | 978 | ||
| AK138228.1 | - | 2 | 1209 | |||
| S80989.1 | - | 2 | 1002 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hmx2| Gene Symbol: Hmx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145998.3 | NP_666110.1 | 2 | 1208 | ||
| GenBank mRNA | AK053014.1 | - | 2 | 978 | ||
| AK138228.1 | - | 2 | 1209 | |||
| S80989.1 | - | 2 | 1002 | |||