siRNA Details
| Gene Symbol: Hmx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145998.3 | NP_666110.1 | 2 | 747 | ||
| GenBank mRNA | AK053014.1 | - | 2 | 517 | ||
| AK138228.1 | - | 2 | 748 | |||
| BC023402.1 | - | 3 | 617 | 5043712 | ||
| S80989.1 | - | 2 | 541 | |||

Hmx2
Genomic Map