siRNA Details
| Gene Symbol: Hn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008258.1 | NP_032284.1 | 2 | 175 | ||
| GenBank mRNA | AK004909.1 | - | 2 | 190 | ||
| AK010057.1 | - | 2 | 190 | |||
| AK013122.1 | - | 2 | 187 | |||
| AK014212.1 | - | 2 | 191 | |||
| AK152560.1 | - | 2 | 192 | |||
| AK159784.1 | - | 2 | 189 | |||
| AK167466.1 | - | 2 | 189 | |||
| AK167961.1 | - | 2 | 179 | |||
| AK168632.1 | - | 2 | 190 | |||
| BC003758.1 | - | 2 | 164 | 3591830 | ||
| U90123.1 | - | 2 | 175 | |||

Hn1
Genomic Map