siRNA Details
| Gene Symbol: Foxa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008260.2 | NP_032286.1 | 2 | 488 | ||
| GenBank mRNA | BC037083.1 | - | 2 | 491 | 5101155 | |
| X74938.1 | - | 2 | 485 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxa3| Gene Symbol: Foxa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008260.2 | NP_032286.1 | 2 | 488 | ||
| GenBank mRNA | BC037083.1 | - | 2 | 491 | 5101155 | |
| X74938.1 | - | 2 | 485 | |||