siRNA Details
| Gene Symbol: HOXA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019102.3 | NP_061975.2 | 2 | 740 | ||
| GenBank mRNA | BC013682.1 | - | 2 | 727 | 3863578 | |
| BI822268.1 | - | 2 | 739 | |||
| BT009744.1 | - | 2 | 679 | |||
| CR542256.1 | - | 2 | 679 | |||
| CR542277.1 | - | 2 | 679 | |||

HOXA5
Genomic Map