siRNA Details
| Gene Symbol: Hnrnpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301340.1 | NP_001288269.1 | 15 | 1454 | ||
| NM_001301341.1 | NP_001288270.1 | 17 | 1581 | |||
| NM_001301343.1 | NP_001288272.1 | 15 | 1440 | |||
| NM_001301344.1 | NP_001288273.1 | 14 | 1382 | |||
| NM_001301345.1 | NP_001288274.1 | 14 | 1382 | |||
| NM_025279.3 | NP_079555.1 | 15 | 1454 | |||
| GenBank mRNA | AK011428.1 | - | 15 | 1459 | ||
| AK051313.1 | - | 14 | 1376 | |||
| AK078777.1 | - | 17 | 1582 | |||
| AK088462.1 | - | 14 | 1369 | |||
| AK128891.1 | - | 15 | 1447 | |||
| AK135519.1 | - | 15 | 1368 | |||
| AK144245.1 | - | 8664 | ||||
| AK150848.1 | - | 15 | 1434 | |||
| AK151689.1 | - | 14 | 1336 | |||
| AK152927.1 | - | 14 | 1336 | |||
| AK160615.1 | - | 15 | 1424 | |||
| AK160887.1 | - | 15 | 1456 | |||
| AK166656.1 | - | 15 | 1459 | |||
| AK166927.1 | - | 15 | 1412 | |||
| AK167600.1 | - | 15 | 1445 | |||
| AK168918.1 | - | 11 | 1357 | |||
| AK169064.1 | - | 15 | 1444 | |||
| AK203272.1 | - | 1 | 83 | |||
| AK204979.1 | - | 1 | 83 | |||
| AK207611.1 | - | 1 | 83 | |||
| AK211652.1 | - | 1 | 83 | |||
| AK215933.1 | - | 1 | 83 | |||
| BC006694.1 | - | 15 | 1421 | 3583037 | ||
| BC089328.1 | - | 15 | 1377 | 6402614 | ||
| EU604547.1 | - | 13 | 1235 | |||

Hnrnpk
Genomic Map