siRNA Details
| Gene Symbol: Hnrnpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177301.5 | NP_796275.3 | 5, 6 | 828 | ||
| GenBank mRNA | AK044748.1 | - | 5, 6 | 604 | ||
| AK085906.1 | - | 5, 6 | 779 | |||
| AK144684.1 | - | 5, 6 | 782 | |||
| BC027206.1 | - | 5, 6 | 805 | 4218471 | ||
| BC099683.1 | - | 5, 6 | 823 | 6530863 | ||

Hnrnpl
Genomic Map