siRNA Details
| Gene Symbol: Hoxa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008265.3 | NP_032291.1 | 2 | 1402 | ||
| GenBank mRNA | AK076098.1 | - | 2 | 1403 | ||
| X13538.1 | - | 1 | 810 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxa4| Gene Symbol: Hoxa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008265.3 | NP_032291.1 | 2 | 1402 | ||
| GenBank mRNA | AK076098.1 | - | 2 | 1403 | ||
| X13538.1 | - | 1 | 810 | |||