siRNA Details
| Gene Symbol: Hoxa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010453.5 | NP_034583.1 | 1 | 302 | ||
| GenBank mRNA | AB221550.1 | - | 1 | 65 | ||
| AK144356.1 | - | 1 | 43 | |||
| BC011063.1 | - | 1 | 126 | 3985274 | ||
| M28021.1 | - | 1 | 302 | |||
| M36604.1 | - | 1 | 179 | |||
| X16840.1 | - | 1 | 179 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008762946, XM_003752180, XM_003749749.

Hoxa5
Genomic Map