siRNA Details
| Gene Symbol: HOXA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024014.3 | NP_076919.1 | 1 | 177 | ||
| GenBank mRNA | BC069497.1 | - | 1 | 152 | 7262260 | |
| BC104915.1 | - | 1 | 177 | 8143918 | ||
| BC104917.1 | - | 1 | 177 | 8143920 | ||

HOXA6
Genomic Map