siRNA Details
| Gene Symbol: Hoxb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010459.7 | NP_034589.3 | 2 | 1957 | ||
| GenBank mRNA | AK133945.1 | - | 2 | 1486 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxb4| Gene Symbol: Hoxb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010459.7 | NP_034589.3 | 2 | 1957 | ||
| GenBank mRNA | AK133945.1 | - | 2 | 1486 | ||