siRNA Details
| Gene Symbol: Hoxb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008268.2 | NP_032294.2 | 2 | 703 | ||
| GenBank mRNA | AK012146.1 | - | 2 | 509 | ||
| AK133512.1 | - | 2 | 619 | |||
| BC040755.1 | - | 2334 | ||||
| BC103595.1 | - | 2 | 623 | 40046040 | ||
| BC103596.1 | - | 2 | 623 | 40046042 | ||
| BC103604.1 | - | 2 | 623 | 40046047 | ||
| BC103607.1 | - | 2 | 623 | 40046044 | ||
| M26283.1 | - | 2 | 703 | |||

Hoxb5
Genomic Map