siRNA Details
| Gene Symbol: HOXA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006896.3 | NP_008827.2 | 2 | 698 | ||
| GenBank mRNA | AF026397.1 | - | 2 | 698 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXA7| Gene Symbol: HOXA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006896.3 | NP_008827.2 | 2 | 698 | ||
| GenBank mRNA | AF026397.1 | - | 2 | 698 | ||