siRNA Details
| Gene Symbol: Hoxd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013555.4 | NP_038583.1 | 2 | 1654 | ||
| GenBank mRNA | AK134427.1 | - | 2 | 1444 | ||
| AK144746.1 | - | 2 | 799 | |||
| BC019150.1 | - | 2 | 1689 | 5064578 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxd9| Gene Symbol: Hoxd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013555.4 | NP_038583.1 | 2 | 1654 | ||
| GenBank mRNA | AK134427.1 | - | 2 | 1444 | ||
| AK144746.1 | - | 2 | 799 | |||
| BC019150.1 | - | 2 | 1689 | 5064578 | ||