siRNA Details
| Gene Symbol: Hpca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130419.2 | NP_001123891.1 | 4 | 1369 | ||
| NM_001286081.1 | NP_001273010.1 | 4 | 1282 | |||
| NM_001286083.1 | NP_001273012.1 | 4 | 1214 | |||
| NM_010471.4 | NP_034601.1 | 4 | 1273 | |||
| GenBank mRNA | AK002992.1 | - | 4 | 1273 | ||
| BC049607.1 | - | 4 | 1238 | 6595363 | ||
| BC058588.1 | - | 3 | 1187 | 5360894 | ||

Hpca
Genomic Map