siRNA Details
| Gene Symbol: Hpgd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008278.2 | NP_032304.2 | 4 | 417 | ||
| GenBank mRNA | AK002472.1 | - | 4 | 385 | ||
| AK146038.1 | - | 4 | 385 | |||
| BC021157.1 | - | 4 | 376 | 4208980 | ||
| U44389.1 | - | 4 | 376 | |||

Hpgd
Genomic Map