siRNA Details
| Gene Symbol: Hprt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013556.2 | NP_038584.2 | 3, 4 | 460 | ||
| GenBank mRNA | AK002286.1 | - | 3, 4 | 405 | ||
| AK082827.1 | - | 3, 4 | 350 | |||
| AK088114.1 | - | 3, 4 | 350 | |||
| AK146626.1 | - | 3, 4 | 406 | |||
| BC004686.1 | - | 3, 4 | 375 | 3500885 | ||
| BC083145.1 | - | 3, 4 | 346 | 30445781 | ||
| J00423.1 | - | 3, 4 | 401 | |||

Hprt
Genomic Map