siRNA Details
| Gene Symbol: Hr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021877.3 | NP_068677.2 | 12 | 3092 | ||
| GenBank mRNA | AB221596.1 | - | 7 | 777 | ||
| AK076280.1 | - | 6 | 577 | |||
| AY312855.1 | - | 11 | 2775 | |||
| AY547390.1 | - | 11 | 2772 | |||
| AY547391.1 | - | 11 | 2772 | |||
| BC049182.1 | - | 12 | 3092 | 6468299 | ||
| Z32675.1 | - | 11 | 2775 | |||

Hr
Genomic Map