siRNA Details
| Gene Symbol: Hrh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008285.3 | NP_032311.2 | 3 | 1435 | ||
| GenBank mRNA | AK046607.1 | - | 3 | 1436 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hrh1| Gene Symbol: Hrh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008285.3 | NP_032311.2 | 3 | 1435 | ||
| GenBank mRNA | AK046607.1 | - | 3 | 1436 | ||
Set Membership: |
|