siRNA Details
| Gene Symbol: Hs3st1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010474.2 | NP_034604.1 | 2 | 1133 | ||
| GenBank mRNA | AF019385.1 | - | 1134 | |||
| AK087753.1 | - | 2 | 1112 | |||
| AK132292.1 | - | 2 | 1144 | |||
| BC009133.1 | - | 2 | 1095 | 3155049 | ||

Hs3st1
Genomic Map