siRNA Details
| Gene Symbol: Hs3st3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178870.5 | NP_849201.1 | 1, 2 | 1293 | ||
| GenBank mRNA | AK032282.1 | - | 1, 2 | 261 | ||
| AK051284.1 | - | 1, 2 | 1294 | |||
| BC094320.1 | - | 1, 2 | 1347 | 6400013 | ||

Hs3st3a1
Genomic Map