siRNA Details
Gene Symbol: Hsd17b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008291.3 | NP_032317.2 | 11 | 944 | ||
GenBank mRNA | BB041573.2 | - | 3 | 325 | ||
U66827.1 | - | 11 | 929 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hsd17b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008291.3 | NP_032317.2 | 11 | 944 | ||
GenBank mRNA | BB041573.2 | - | 3 | 325 | ||
U66827.1 | - | 11 | 929 |