siRNA Details
| Gene Symbol: HPCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002143.2 | NP_002134.2 | 2 | 173 | ||
| XM_005270792.2 | XP_005270849.1 | 2 | 309 | |||
| XM_017001118.1 | XP_016856607.1 | 2 | 645 | |||
| GenBank mRNA | AK313907.1 | - | 2 | 177 | ||
| BC001777.1 | - | 2 | 173 | 3355354 | ||
| BT019750.1 | - | 1 | 133 | |||
| BT019789.1 | - | 1 | 133 | |||
| D16593.1 | - | 1 | 143 | |||

HPCA
Genomic Map