siRNA Details
| Gene Symbol: Hsd17b7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010476.3 | NP_034606.3 | 8 | 908 | ||
| GenBank mRNA | AK028380.1 | - | 8 | 864 | ||
| AK050211.1 | - | 8 | 865 | |||
| AK077914.1 | - | 8 | 906 | |||
| BC011464.1 | - | 8 | 872 | |||
| Y15733.1 | - | 8 | 898 | |||

Hsd17b7
Genomic Map