siRNA Details
Gene Symbol: Hsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008296.2 | NP_032322.1 | 2 | 292 | ||
GenBank mRNA | AK075651.1 | - | 2 | 282 | ||
AK082507.1 | - | 316 | ||||
AK089144.1 | - | 2 | 317 | |||
BC013716.1 | - | 2 | 292 | 3978881 | ||
BC094064.1 | - | 2 | 302 | 4158683 | ||
KM043052.1 | - | 2 | 292 | |||
KM043053.1 | - | 2 | 292 | |||
KM043054.1 | - | 2 | 292 | |||
KM043055.1 | - | 2 | 292 | |||
X61753.1 | - | 2 | 275 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
