siRNA Details
Gene Symbol: Hspg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008305.3 | NP_032331.2 | 10 | 1252 | ||
GenBank mRNA | AK147294.1 | - | 10 | 1253 | ||
AK147396.1 | - | 10 | 1253 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hspg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008305.3 | NP_032331.2 | 10 | 1252 | ||
GenBank mRNA | AK147294.1 | - | 10 | 1253 | ||
AK147396.1 | - | 10 | 1253 |