siRNA Details
| Gene Symbol: Ndst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008306.4 | NP_032332.2 | 1 | 265 | ||
| GenBank mRNA | AF049894.1 | - | 1 | 111 | ||
| AF074926.1 | - | 1 | 111 | |||
| AK035642.1 | - | 2 | 674 | |||
| AK147282.1 | - | 2 | 658 | |||
| AK156504.1 | - | 2 | 618 | |||
| AK170041.1 | - | 2 | 591 | |||
| AK171013.1 | - | 2 | 591 | |||
| BC066098.1 | - | 1 | 265 | 30533342 | ||
| BC079561.1 | - | 1 | 265 | 30533342 | ||

Ndst1
Genomic Map