siRNA Details
| Gene Symbol: HOXB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002145.3 | NP_002136.1 | 2 | 571 | ||
| XM_005257275.4 | XP_005257332.1 | 2 | 140 | |||
| GenBank mRNA | BC074805.2 | - | 2 | 471 | 30915528 | |
| BC074806.2 | - | 2 | 471 | 30915314 | ||
| M73999.1 | - | 2 | 195 | |||
| X14571.1 | - | 1 | 25 | |||
| X16665.1 | - | 2 | 529 | |||

HOXB2
Genomic Map