siRNA Details
| Gene Symbol: Htr1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285482.1 | NP_001272411.1 | 3 | 1612 | ||
| NM_001285483.1 | NP_001272412.1 | 3 | 1434 | |||
| NM_001285484.1 | NP_001272413.1 | 2 | 1341 | |||
| NM_008309.5 | NP_032335.2 | 2 | 1454 | |||
| GenBank mRNA | AK082016.1 | - | 2 | 1458 | ||
| BC103532.1 | - | 2 | 1465 | 40039729 | ||
| BC103535.1 | - | 3 | 1428 | 40039732 | ||
| L20335.1 | - | 1 | 85 | |||
More Information
Set Membership: |
|

Htr1d
Genomic Map