siRNA Details
| Gene Symbol: Htr5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008314.2 | NP_032340.2 | 1 | 1059 | ||
| GenBank mRNA | AK141636.1 | - | 1 | 858 | ||
| BC115911.1 | - | 1 | 751 | 40104765 | ||
| BC116654.1 | - | 1 | 751 | 40104769 | ||
More Information
Set Membership: |
|

Htr5a
Genomic Map