siRNA Details
| Gene Symbol: Htr5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC114988.1 | - | 1 | 853 | 40056207 | |
| BC115825.1 | - | 1 | 853 | 40056204 | ||
| X69867.1 | - | 1 | 999 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Htr5b| Gene Symbol: Htr5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC114988.1 | - | 1 | 853 | 40056207 | |
| BC115825.1 | - | 1 | 853 | 40056204 | ||
| X69867.1 | - | 1 | 999 | |||