siRNA Details
| Gene Symbol: Htr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021358.2 | NP_067333.1 | 1 | 543 | ||
| GenBank mRNA | AF134158.1 | - | 1 | 543 | ||
| AK134977.1 | - | 2 | 1222 | |||
| BC116652.1 | - | 1 | 543 | 40104888 | ||
| BC116653.1 | - | 1 | 543 | 40104889 | ||
More Information
Set Membership: |
|

Htr6
Genomic Map