siRNA Details
| Gene Symbol: Elavl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010485.3 | NP_034615.2 | 5 | 734 | ||
| GenBank mRNA | AK034536.1 | - | 5 | 721 | ||
| AK075818.1 | - | 5 | 696 | |||
| AK078333.1 | - | 5 | 1050 | |||
| AK080365.1 | - | 5 | 692 | |||
| AK089088.1 | - | 4 | 441 | |||
| AK148528.1 | - | 5 | 662 | |||
| AK161667.1 | - | 4 | 451 | |||
| AK183003.1 | - | 1 | 47 | |||
| AK186913.1 | - | 1 | 47 | |||
| AK201115.1 | - | 3 | 253 | |||
| AK207077.1 | - | 1 | 47 | |||
| AK214281.1 | - | 3 | 279 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001108848, AB212679, BC168972.

Elavl1
Genomic Map