siRNA Details
Gene Symbol: Iapp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010491.2 | NP_034621.1 | 3 | 447 | ||
GenBank mRNA | BC027527.1 | - | 3 | 418 | 1379049 | |
M25389.1 | - | 2 | 354 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Iapp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010491.2 | NP_034621.1 | 3 | 447 | ||
GenBank mRNA | BC027527.1 | - | 3 | 418 | 1379049 | |
M25389.1 | - | 2 | 354 |